missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-23 p19 Monoclonal Antibody (G23-8), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£2110.00 - £12950.00
Specifications
| Antigen | IL-23 p19 |
|---|---|
| Clone | G23-8 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Functional Assay, Neutralization, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15537816
|
Invitrogen™
16-7232-38 |
5 mg |
£2110.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17141512
|
Invitrogen™
16-7232-025 |
25 mg |
£8700.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17151512
|
Affymetrix eBioscience
16-7232-050 |
50 mg | |||||||
Description
IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-γ. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-γ production by naive and memory T cells, as compared to IL-12.Specifications
| IL-23 p19 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 83430 | |
| Primary | |
| 4°C | |
| IL23A |
| G23-8 | |
| ELISA, Functional Assay, Neutralization, Western Blot | |
| Functional Grade | |
| Rat | |
| Mouse | |
| Q9EQ14 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title