Learn More
Invitrogen™ IL-23 p19 Antibody Cocktail, eBioscience™
Antibody Cocktail
£467.00
Specifications
| Antigen | IL-23 p19 |
|---|---|
| Clone | eBio473P19 (eBio11P19, eBio25P19), eBio25P19) |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | ELISA, Western Blot |
Description
Description: The antibody eBio473P19 reacts with the p19 subunit of human IL-23. The eBio473P19 antibody was generated from immunization with authentic, insect cell-expressed, recombinant human IL-23 heterodimer. The use of a p19-specific capture antibody and a p40-specific detection antibody yields a human IL-23 sandwich ELISA exquisitely specific for human IL-23. IL-12 p40 monomer and IL-12 p70 were run in the assay at 200 ng/mL with no interference or cross-reactivity observed. A panel of 20 unrelated cytokines was also run in the IL-23 ELISA at 100 ng/mL with no cross reactivity observed. The assay has been validated by specific detection of significant levels of native human IL-23 protein in supernatants from a variety of different activated dendritic cell populations. IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-g. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells ...
IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-γ. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-γ production by naive and memory T cells, as compared to IL-12.Specifications
| IL-23 p19 | |
| 0.5 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9NPF7 | |
| 51561 | |
| Primary | |
| Affinity chromatography |
| eBio473P19 (eBio11P19, eBio25P19), eBio25P19) | |
| 4°C | |
| Cocktail | |
| Liquid | |
| IgG1 κ | |
| Human | |
| IL23A | |
| il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 | |
| IL23A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.