Learn More
IL-23 p19, Clone: eBio473P19 (eBio11P19, eBio25P19), Antibody Cocktail, eBioscience™
Antibody Cocktail
Brand: Invitrogen 14-7238-80
Description
This gene encodes a subunit of a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. The cytokine is a disulfide-linked heterodimer composed of the 35-kD subunit encoded by this gene, and a 40-kD subunit that is a member of the cytokine receptor family. This cytokine is required for the T-cell-independent induction of interferon (IFN)-gamma, and is important for the differentiation of both Th1 and Th2 cells. The responses of lymphocytes to this cytokine are mediated by the activator of transcription protein STAT4. Nitric oxide synthase 2A (NOS2A/NOS2) is found to be required for the signaling process of this cytokine in innate immunity.
Description: The antibody eBio473P19 reacts with the p19 subunit of human IL-23. The eBio473P19 antibody was generated from immunization with authentic, insect cell-expressed, recombinant human IL-23 heterodimer. The use of a p19-specific capture antibody and a p40-specific detection antibody yields a human IL-23 sandwich ELISA exquisitely specific for human IL-23. IL-12 p40 monomer and IL-12 p70 were run in the assay at 200 ng/ml with no interference or cross-reactivity observed. A panel of 20 unrelated cytokines was also run in the IL-23 ELISA at 100 ng/ml with no cross reactivity observed. The assay has been validated by specific detection of significant levels of native human IL-23 protein in supernatants from a variety of different activated dendritic cell populations.IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-g. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-g production by naive and memory T cells, as compared to IL-12. IL-23-dependent, IL-17-producing CD4+ T cells (Th-17 cells) have been identified as a unique subset of Th cells that develops along a pathway that is distinct from the Th1- and Th2- cell differentiation pathways. The hallmark effector molecules of Th1 and Th2 cells, e.g., IFN-g and IL-4, have each been found to negatively regulate the generation of these Th-17 cells. More recently, de novo differentiation of Th-17 cells in the absence of IL-23 has been demonstrated by treatment of naive CD4 cells with TGF beta1 and IL-6.Applications Reported: This eBio473P19 antibody has been reported for use in immunoblotting (WB), and ELISA.Applications Tested: The eBio473P19 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human IL-23 (p19p40) protein levels in combination with the biotinylated (p40-specific) C8.6 antibody (13-7129) for detection and recombinant human IL-23 (14-8239) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 2.5-8.0μg/mL. A standard curve consisting of doubling dilutions of the recombinant IL-23 standard over the range of 2000 pg/mL - 15 pg/mL should be included in each ELISA plate. TMB, rather than ABTS, should be used as the substrate.The antibody eBio473p19 also works in immunoblotting under both reducing and nonreducing conditions at 2μg/mL.*eBio473P19 is a cocktail of the two clones, eBio11P19 and eBio25P19 at 0.5mg/mL each. Please contact Tech Support for more information regarding the concentration.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications
| IL-23 p19 | |
| Cocktail | |
| 0.5mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9NPF7 | |
| IL23A | |
| Affinity chromatography | |
| RUO | |
| 51561 | |
| Store at 2-8°C. | |
| Liquid |
| ELISA, Western Blot | |
| eBio473P19 | |
| Unconjugated | |
| IL23A | |
| IL-23, IL-23A, IL23P19, MGC79388, P19, SGRF | |
| Mouse | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.