missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22RA2 Rabbit anti-Human, CoraLite 650, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL650-22861-100UL
This item is not returnable.
View return policy
Description
IL-22RA2 Polyclonal antibody apecifically detetects IL-22RA2 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| IL-22RA2 | |
| Polyclonal | |
| CoraLite 650 | |
| BC125167 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IL22RA2 | |
| IL-22RA2 fusion protein Ag18854 | |
| 100 μL | |
| 116379 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction