missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22BP/IL22 RA2 Antibody (ARGX-112), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28505CL1
This item is not returnable.
View return policy
Description
IL-22BP/IL22 RA2 Monoclonal antibody specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-22BP/IL22 RA2 | |
| Monoclonal | |
| CoraFluor 1 | |
| class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
| IL-22Ra | |
| 0.1 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 116379 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ARGX-112 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction