Learn More
Invitrogen™ IL-22 Receptor alpha Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5144626
Product Code. 17920773
  • £425.00 / 100µL
Description
Positive control: 293T cell lysate, HepG2 cell lysate, K562 cell lysate, Hela, SiHa, mouse kidney tissue, rat pancreas tissue. Predicted band size: 63 kDa Subcellular Location: Membrane.
A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-22 Receptor alpha
Polyclonal
Unconjugated
Il22ra1
9130219A07Rik; CRF2-9; Cytokine receptor class-II member 9; cytokine receptor family 2 member 9; ENSMUSG00000073746; il-22 receptor; IL-22 receptor subunit alpha-1; IL22R; IL-22R; IL22R1; IL22RA1; IL-22RA1; IL-22R-alpha-1; ILN; Interleukin; interleukin 22 receptor subunit alpha 1; interleukin 22 receptor, alpha 1; interleukin-22 receptor alpha chain; interleukin-22 receptor subunit alpha-1; RGD1559655; zcytoR11
Rabbit
Affinity chromatography
RUO
230828, 362629, 58985
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q80XZ4, Q8N6P7
Il22ra1
Synthetic peptide within human IL-22RA1 aa 1-50/574.
100 μL
Primary
Human, Mouse, Rat