missing translation for 'onlineSavingsMsg'
Learn More

IL-21R Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18431617 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431617 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431617 Supplier Novus Biologicals Supplier No. NBP176739MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL-21R Polyclonal antibody specifically detects IL-21R in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-21R
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD360 antigen, IL-21 receptor, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to amino acids 97 to 111 of human IL-21 receptor precursor. The immunogen is located within amino acids 80 - 130 of IL-21 Receptor. Amino Acid Squence: NITDQSGNYSQECGS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 50615
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.