Learn More
Invitrogen™ IL-21 Polyclonal Antibody

Description
Suggested positive controls are Human Tonsil, Mouse Spleen, or Rat Spleen Lysate. PA5-19981 can be used with blocking peptide PEP-0105. Predicted species reactivity based on immunogen sequence: Bovine: (100%), Pig: (87%). At least two isoforms of IL-21 are known to exist; this antibody will detect both isoforms. IL-21 antibody is predicted to not cross-react with other members of the interleukin protein family.
Specifications
Specifications
| Antigen | IL-21 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | IL21 |
| Gene Accession No. | A3QPB9, Q9ES17, Q9HBE4 |
| Gene Alias | CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11 |
| Gene Symbols | IL21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.