Learn More
Invitrogen™ IL-21 Monoclonal Antibody (FFA21), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 12-7211-80
Description
Description: This FFA21 monoclonal antibody reacts with mouse interleukin-21 (IL-21). IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by NKT, T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1, IL-4 for Th2 and IL-17A for Th17, IL-21 is critical for TFH cell effector function. This cytokine plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintece by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. High levels of IL-21 are present in chemically-induced colitis models. IL-21-deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. Preliminary data suggest that clone FFA21 recognizes a different epitope than anti-mouse IL-21 clone mhalx21 (cat. 51-7213). Applications Reported: This FFA21 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
Specifications
| IL-21 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9ES17 | |
| IL21 | |
| Affinity chromatography | |
| RUO | |
| 60505 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| FFA21 | |
| PE | |
| IL21 | |
| CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11 | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.