missing translation for 'onlineSavingsMsg'
Learn More

IL-20R alpha Antibody (MM0394-9F40), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18420426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420426 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420426 Supplier Novus Biologicals Supplier No. NBP211695AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20R alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0394-9F40
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
Host Species Mouse
Immunogen Human recombinant IL-20R alpha (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.