missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-20R alpha Antibody (Cheng Kung U. patent anti-IL-20R1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28332-1mg
This item is not returnable.
View return policy
Description
IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-20R alpha | |
| Monoclonal | |
| Unconjugated | |
| Q9UHF4 | |
| Human | |
| Protein A purified | |
| RUO | |
| 53832 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Cheng Kung U. patent anti-IL-20R1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
| IL-20Ra | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur