missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20R alpha Antibody (Cheng Kung U. patent anti-IL-20R1), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28332CL1
This item is not returnable.
View return policy
Description
IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-20R alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
| IL-20Ra | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Cheng Kung U. patent anti-IL-20R1 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| 53832 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction