missing translation for 'onlineSavingsMsg'
Learn More

IL-20R alpha Antibody (038), DyLight 755, Novus Biologicals™

Product Code. 18389750 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389750 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389750 Supplier Novus Biologicals Supplier No. NBP289469IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20R alpha
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 38
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-20R alpha (NP_055247.3; Met 1-Lys 250).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Test Specificity No cross-reactivity in ELISA with: Human IFNgR1 Human IL10RB
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.