missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114751
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Interleukin 20.
Cytokines that signal through Class II receptors form a distinct family that includes the interferons and interleukin 10 (IL-10). Recent identification of several IL-10 homologs has defined a cytokine subfamily that includes AK155, IL-19, IL-20, IL-22, and IL-24. IL-20 belongs to the IL-10 family and plays a role in skin inflammation and the development of hematopoietic cells. Little is known about its other biological functions and clinical implications. These cytokines display pro-inflammatory activities in different tissues, including skin.
Specifications
| IL-20 | |
| Polyclonal | |
| Unconjugated | |
| Il20 | |
| cytokine Zcyto10; four alpha helix cytokine; four alpha helix cytokine ZCYTO10; four helix bundle cytokine 10; il 20; IL10D; Il20; IL-20; ILN; Interleukin; interleukin 20; interleukin-20; MGC96907; PRO1801; UNQ852; UNQ852/PRO1801; ZCYTO10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50604, 58181 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9JKV9, Q9NYY1 | |
| Il20 | |
| A synthesized peptide derived from human IL20(Accession Q9NYY1), corresponding to amino acid residues E41-L91. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction