missing translation for 'onlineSavingsMsg'
Learn More

IL-20 R beta/FNDC6 Antibody (20RNTC), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18425083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425083 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425083 Supplier Novus Biologicals Supplier No. NBP200466MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-20 R beta/FNDC6 Monoclonal antibody specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20 R beta/FNDC6
Applications Flow Cytometry
Classification Monoclonal
Clone 20RNTC
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923
Host Species Rat
Immunogen The immunogen for this antibody is FNDC6 (IL-20R2).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53833
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.