Learn More
IL-20 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen 500-P190GBT-1MG
Description
AA Sequence of recombinant protein: MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-20. Anti-Human IL-20-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIL-20 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-20 (500-P190G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIL-20. Western Blot: To detect Human IL-20 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-20 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P190GBT-1MG will be provided as 2 x 500 μg
Cytokines that signal through Class II receptors form a distinct family that includes the interferons and interleukin 10 (IL-10). Recent identification of several IL-10 homologs has defined a cytokine subfamily that includes AK155, IL-19, IL-20, IL-22, and IL-24. IL-20 belongs to the IL-10 family and plays a role in skin inflammation and the development of hematopoietic cells. Little is known about its other biological functions and clinical implications. These cytokines display pro-inflammatory activities in different tissues, including skin.
Specifications
| IL-20 | |
| Polyclonal | |
| Biotin | |
| Il20 | |
| cytokine Zcyto10; four alpha helix cytokine; four alpha helix cytokine ZCYTO10; four helix bundle cytokine 10; il 20; IL10D; Il20; IL-20; ILN; Interleukin; interleukin 20; interleukin-20; MGC96907; PRO1801; UNQ852; UNQ852/PRO1801; ZCYTO10 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 50604 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9NYY1 | |
| Il20 | |
| E.coli-derived Recombinant Human IL-20 | |
| 2 x 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.