missing translation for 'onlineSavingsMsg'
Learn More

IL-20 Antibody (287023) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30735504 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735504 Supplier Novus Biologicals Supplier No. FAB11021MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-20 Monoclonal antibody specifically detects IL-20 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20
Applications Western Blot
Classification Monoclonal
Clone 287023
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
Host Species Rat
Immunogen E. coli-derived recombinant human IL-20, Leu25-Glu176, Accession # AAF36679
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50604
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.