missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-20 Antibody (03) - Azide and BSA Free, Novus Biologicals™

Product Code. 30043399 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30043399 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30043399 Supplier Novus Biologicals™ Supplier No. NBP326912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-20 Monoclonal antibody specifically detects IL-20 in Human samples. It is validated for ELISA,Sandwich ELISA Detection
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20
Applications ELISA, Sandwich ELISA Detection
Classification Monoclonal
Clone 03
Conjugate Unconjugated
Dilution ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 μm filtered solution in PBS
Gene Alias Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-20 (Uniprot#: Q9NYY1; Leu25-Glu176).
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50604
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.