Learn More
Invitrogen™ IL-2 Monoclonal Antibody (JES6-1A12), eBioscience™, Invitrogen™

Rat Monoclonal Antibody
Brand: Invitrogen™ 14-7022-81
Description
Description: The JES6-1A12 antibody reacts with mouse interleukin-2 (IL-2). The JES6-1A12 antibody is a neutralizing antibody. Mouse IL-2 (also known as TCGF) is a 17 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK cells, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions, and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active. Applications Reported: The JES6-1A12 antibody has been reported for use in ELISA capture, ELISPOT capture, neutralization of IL-2 bioactivity, IP, and intracellular staining. Applications Tested: The JES6-1A12 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse Interleukin-2 (IL-2) in combination with the biotin JES6-5H4 (13-7021) antibody for detection and recombit mouse IL-2 (14-8021) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 500 pg/mL - 4 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
| IL-2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P04351 | |
| IL2 | |
| Affinity chromatography | |
| RUO | |
| 16183 | |
| 4°C | |
| Liquid |
| Immunoprecipitation, Immunostaining | |
| JES6-1A12 | |
| Unconjugated | |
| IL2 | |
| aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
| Rat | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.