Learn More
IL-2 Rat anti-Human, PE, Clone: MQ1-17H12, BD
Rat Monoclonal Antibody
£59.66 - £238.64
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | MQ1-17H12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15860579
|
BD Biosciences
560902 |
25 Tests |
£62.10 £59.66 / Each Save £2.44 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15809718
|
BD Biosciences
559334 |
100 Tests |
£249.00 £238.64 / Each Save £10.36 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MQ1-17H12 monoclonal antibody specifically binds to the multifunctional cytokine, human Interleukin-2 (IL-2). IL-2 is produced by activated T cells and has multiple functions that can affect the growth, proliferation, differentiation and survival of many different target cell types including T cells, B cells, NK cells, monocytes and macrophages. The immunogen used to generate the MQ1-17H12 hybridoma was purified recombinant human IL-2 protein. The MQ1-17H12 antibody reportedly neutralizes the biological activity of human IL-2.
Specifications
| IL-2 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| MQ1-17H12 | |
| Monoclonal | |
| Rat | |
| Human | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.