Learn More
Invitrogen™ IL-2 Monoclonal Antibody (MQ1-17H12), eBioscience™, Invitrogen™

Description
Description: The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody. Applications Reported: The MQ1-17H12 antibody has been reported for use as capture antibody for ELISA and for intracellular staining for flow cytometric analysis. Fluorochrome conjugated MQ1-17H12 antibody is recommended for use in intracellular staining for flow cytometry. Applications Tested: The MQ1-17H12 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human Interleukin-2 (IL-2) in combination with the biotin anti-human IL-2 poly (13-7028) antibody for detection and recombinant human IL-2 (14-8029) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | IL-2 |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | MQ1-17H12 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | IL2 |
| Gene Accession No. | P60568 |
| Gene Alias | aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.