missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30199-1-AP-20UL
This item is not returnable.
View return policy
Description
The IL-2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of rat IL-2. This antibody recognizes rat antigen. The IL-2 antibody has been validated for the following applications: ELISA, FC (Intra) analysis.Specifications
| IL-2 | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Rat | |
| Polyclonal Antibody | |
| IgG |
| ELISA, Flow Cytometry (Intracellular Staining) | |
| 500 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 116562 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction