missing translation for 'onlineSavingsMsg'
Learn More

IL-2, Rabbit anti-Human, Clone: LNKB-2, Novus Biologicals™

Product Code. 18315021 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18315021 0.025 mg 25µg
18365640 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18315021 Supplier Novus Biologicals Supplier No. NBP2810470.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications Western Blot, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone LNKB-2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias aldesleukin, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Gene Symbols IL2
Host Species Rabbit
Immunogen Recombinant monoclonal antibody to IL-2.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3558
Test Specificity Recognises IL-2.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.