missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 R beta Antibody (TM-b1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£157.00 - £489.00
Specifications
| Antigen | IL-2 R beta |
|---|---|
| Clone | TM-b1 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess |
| Applications | Flow Cytometry, Immunoprecipitation, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280592
|
Novus Biologicals
NBP1-43355-0.025MG |
0.025 mg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18285154
|
Novus Biologicals
NBP1-43355-0.1MG |
0.1 mg |
£375.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740594
|
Novus Biologicals
NBP1-43355-0.5mg |
0.5 mg |
£489.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Block/Neutralize.Specifications
| IL-2 R beta | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunoprecipitation, Blocking Assay | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 3560 | |
| The immunogen for this antibody was Purified CD122. | |
| Primary | |
| Store at 4C. Do not freeze. |
| TM-b1 | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess | |
| Monoclonal | |
| Rat | |
| Mouse | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| IL2RB | |
| IgG2b | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title