missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 R beta Antibody (130122) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5891JF585
This item is not returnable.
View return policy
Description
IL-2 R beta Monoclonal antibody specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| IL-2 R beta | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-2 Rβ, Ala27-Glu240, Accession # P16297 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 130122 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.5 mL | |
| Immunology | |
| 3560 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction