Learn More
PeproTech IL-2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P274BT-500UG
Description
AA Sequence of recombinant protein: MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAIAQSI ISTM. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-2. Anti-Rat IL-2-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-2 (500-P274) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-2. Western Blot: To detect Rat IL-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat IL-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P274BT-1MG will be provided as 2 x 500 μg
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
| IL-2 | |
| Polyclonal | |
| Biotin | |
| IL2 | |
| aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116562 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P17108 | |
| IL2 | |
| E.coli-derived Recombinant Rat IL-2. | |
| 500 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.