missing translation for 'onlineSavingsMsg'
Learn More

IL-2 Mouse anti-Human, Clone: B-G5, Novus Biologicals™

Product Code. 18321527 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321527 500 μg 500µL
18376976 200 μg 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321527 Supplier Novus Biologicals Supplier No. NBP314593500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Functional Assay, ELISPOT
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications Western Blot, Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone B-G5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Functional, ELISPOT
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Host Species Mouse
Immunogen Natural human IL-2
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3558
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.