missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Rabbit anti-Human, Clone: 5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89195-100UL
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| A synthetic peptide corresponding to the center region of the human IL-1ra/IL-1F3/IL1RN | |
| 100 μg | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3557 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction