missing translation for 'onlineSavingsMsg'
Learn More

IL-1ra/IL-1F3/IL1RN Antibody (005), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18375460 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375460 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375460 Supplier Novus Biologicals Supplier No. NBP289195JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1ra/IL-1F3/IL1RN
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human IL-1ra/IL-1F3/IL1RN.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3557
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.