missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-19 Rabbit anti-Human, Clone: 19, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90169-100UL
This item is not returnable.
View return policy
Description
IL-19 Monoclonal specifically detects IL-19 in Human samples. It is validated for ELISA.
Specifications
| IL-19 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 19 | |
| ELISA 1:5000-1:10000 | |
| IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19 | |
| Recombinant Human IL-19 Protein | |
| 100 μg | |
| Biologically Active Proteins, Cytokine Research | |
| 29949 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction