missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-19 Monoclonal Antibody (RN19), eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

£86.75 - £243.00

Specifications

Antigen IL-19
Clone RN19
Concentration 0.5 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15298037
GREENER_CHOICE
View Documents
Invitrogen™
14-7191-81
50 μg
£86.75
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15208047
View Documents
Invitrogen™
14-7191-85
500 μg
£243.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The RN19 antibody reacts with mouse interleukin-19 (IL-19), a member of the IL-10 family, which is secreted by activated keratinocytes and monocytes. Applications Reported: The RN19 antibody has been reported for use in ELISA capture of mouse IL-19. Applications Tested: The RN19 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse Interleukin-19 (IL-19) in combination with a biotinylated polyclonal anti-mouse IL-19 antibody for detection. A suitable range of concentrations of this antibody for ELISA capture is 1.0-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 4000 pg/mL - 30 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
Specifications

Specifications

IL-19
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
il 19; IL-10C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG.1; ZMDA1
IL19
Primary
4°C
IL19
RN19
ELISA
Unconjugated
Rat
Mouse
Q8CJ70
329244
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.