missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-19 Antibody Pair [HRP]

Product Code. 18633640 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18633640

Brand: Novus Biologicals™ NBP2794085Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human IL-19.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human IL-19.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19
Gene ID (Entrez) 29949
Gene Symbol IL19
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 30 ng of recombinant Human IL-19.
  • Capture Antibody: 1 mg/mL of mouse anti-Human IL-19 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human IL-19 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins, Cytokine Research
    Storage Requirements Storage is content dependent.
    Target IL-19
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.