missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18R alpha/IL-1 R5 Antibody (70625), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB840APCCY7
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-18R alpha/IL-1 R5 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8809 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 70625 | |
| Flow Cytometry | |
| CD218a;CDw218a;IL18R1;IL18RA;IL-18Ralpha;IL-18R-alpha;IL18Ralpha2;IL1RRP;IL-1Rrp;Interleukin-18 receptor 1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5, Glu20-Arg329, Accession # Q13478 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction