missing translation for 'onlineSavingsMsg'
Learn More

IL-18R alpha/IL-1 R5 Antibody (112624) [DyLight 488], Novus Biologicals™

Product Code. 30731650 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731650 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731650 Supplier Novus Biologicals Supplier No. FAB12161K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-18R alpha/IL-1 R5
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 112624
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CD218a;CDw218a;IL18R1;IL18RA;IL-18Ralpha;IL-18R-alpha;IL18Ralpha2;IL1RRP;IL-1Rrp;Interleukin-18 receptor 1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha , Ser19-Gly326, Accession # Q61098
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.