missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18 R beta/IL-1 R7/ACPL Antibody (132029), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB118L
This item is not returnable.
View return policy
Description
IL-18 R beta/IL-1 R7/ACPL Monoclonal antibody specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| IL-18 R beta/IL-1 R7/ACPL | |
| Monoclonal | |
| DyLight 550 | |
| Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18RAcP, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 R beta/IL-1 R7, Met1-Arg356, Accession # O95256 | |
| 0.5 mL | |
| CD Markers | |
| 8807 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 132029 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction