missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580719
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL-18.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
Specifications
| IL-18 | |
| Polyclonal | |
| Unconjugated | |
| IL18 | |
| cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 3606 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 5 mg/mL | |
| PBS with no preservative | |
| Q14116 | |
| IL18 | |
| Recombinant Human IL-18 Protein (Met1-Asp193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction