missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18 Recombinant Rabbit Monoclonal Antibody (HL1859), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA547203
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.Specifications
IL-18 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P70380, Q14116 | |
IL18 | |
Recombinant fragment of mouse IL18 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
HL1859 | |
Unconjugated | |
IL18 | |
cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320 | |
Rabbit | |
Protein A | |
RUO | |
16173, 3606 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |