missing translation for 'onlineSavingsMsg'
Learn More

IL-18/IL-1F4 Antibody (314811) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30733058 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733058 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733058 Supplier Novus Biologicals Supplier No. FAB29241MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Canine samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-18/IL-1F4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 314811
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Host Species Mouse
Immunogen E. coli-derived recombinant canine IL-18/IL-1F4, Tyr37-Ser193, Accession # Q9XSR0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3606
Target Species Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.