missing translation for 'onlineSavingsMsg'
Learn More

IL-18/IL-1F4 Antibody (167), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18361501 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361501 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361501 Supplier Novus Biologicals Supplier No. NBP290394AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-18/IL-1F4
Applications ELISA
Classification Monoclonal
Clone 167
Conjugate Alexa Fluor 594
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-18/IL-1F4 (P70380; Asn36-Ser192).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity No cross-reactivity in ELISA with: Human IL1F7 Human IL1F9
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.