missing translation for 'onlineSavingsMsg'
Learn More

IL-18/IL-1F4 Antibody (125-2H), DyLight 350, Novus Biologicals™

Product Code. 18340871 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340871 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340871 Supplier Novus Biologicals Supplier No. NBP281040UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-18/IL-1F4
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 125-2H
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Gene Symbols IL18
Host Species Rabbit
Immunogen Fusion of splenocytes from mice immunized with human IL-18/IL-1F4, with SP2/O mouse myeloma cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity Recognizes the active form of human IL-18/IL-1F4.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.