missing translation for 'onlineSavingsMsg'
Learn More

IL-18 BPa/IL18BP Antibody (224), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18408076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408076 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408076 Supplier Novus Biologicals Supplier No. NBP289445MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-18 BPa/IL18BP Monoclonal antibody specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-18 BPa/IL18BP
Applications ELISA
Classification Monoclonal
Clone 224
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 BPa/IL18BP (Accession#: NP_001034748.1; Met1-Gly194).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10068
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.