missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RD/SEF Rabbit anti-Human, Clone: 10, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | IL-17RD/SEF |
|---|---|
| Clone | 10 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18278336
|
Novus Biologicals
NBP2-89541-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203678
|
Novus Biologicals
NBP2-89541-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17RD/SEF Monoclonal specifically detects IL-17RD/SEF in Human samples. It is validated for ELISA.Specifications
| IL-17RD/SEF | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 54756 | |
| IgG | |
| Protein A purified |
| 10 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef | |
| Recombinant Human IL-17RD/SEF protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title