missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RB Antibody (938951), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB12074B
This item is not returnable.
View return policy
Description
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-17RB | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| IL17RB | |
| Partial recombinant human IL-17RB protein (aa18-289) | |
| 100 μL | |
| Cytokine Research | |
| 55540 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 938951 | |
| Flow Cytometry | |
| CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, EVI27IL-17B receptor, IL-17 receptor B, IL17BRinterleukin 17B receptor, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction