Learn More
IL-17RA/IL-17R Antibody (CL10283) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21174C
Product Code. 30308594
  • £558.00 / 0.10mL
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17RA/IL-17R
Monoclonal
DyLight 650
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
This antibody was developed using a synthetic peptide derived from Q96F46, with the exact immunogen sequence remaining proprietary.
0.1 mL
Innate Immunity
23765
Store at 4⪚C in the dark.
IgG2b
Western Blot
CL10283
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified