missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17R Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHIL17R

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15435033

  • £554.00 / 1 set
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The Human Interleukin 17 Receptor (Hu IL-17) ELISA quantitates Hu IL-17 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-17. Principle of the method The Human IL-17 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
Specifications

Specifications

Q96F46
10 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
23765
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Inflammation
2°C to 8°C
Human
4 hr. 45 min.
12.35-3000 pg/mL
10pg/mL to 3000pg/mL
Biotin
Cytokines & Receptors
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-17, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
IL17RA
<10%
HRP
RUO
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
CANDF5, CD217, CDw217, IL-17RA, IL17R, hIL-17R
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers