Learn More
IL-17F Monoclonal Antibody (SHLR17), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£78.88 - £254.00
Specifications
Antigen | IL-17F |
---|---|
Clone | SHLR17 |
Concentration | 5μL (0.25μg)/test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589806
|
Affymetrix eBioscience
48-7169-42 |
100 Tests |
£254.00
100 tests |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15579806
|
Affymetrix eBioscience
48-7169-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Interleukin 17F is a cytokine belonging to the IL-17 family,that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflamation by increasing chemokine production.
IL-17F (Interleukin 17F, CTLA-8)) is a cytokine belonging to the IL-17 family that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflammation by increasing chemokine production. IL-17F also stimulates induction of other pro-inflammatory cytokines TNF alpha, IL-1 beta, IL-6, and IL-8, and reports strongly suggest the involvement of IL-17 in several chronic inflammatory diseases such as rheumatoid arthritis, psoriasis and multiple sclerosis. TGF-beta (differentiation) and IL-23 (expansion) are required for induction and maintenance of Th17 (IL-17 producing) cells, which in turn induce the other pro-inflammatory cytokines. IL-17F is produced, and exists, as a homo-dimer, with homology to a herpes virus early protein, is one of the six members (IL-17A-F) of this cytokine family, and is well characterized and highly expressed by activated effector memory T cells. IL-17F has been found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1.Specifications
IL-17F | |
5μL (0.25μg)/test | |
Monoclonal | |
Rat | |
Human | |
Interleukin-17F | |
Primary |
SHLR17 | |
Flow Cytometry | |
eFluor 450 | |
RUO | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
IgG1 κ | |
Store at 2-8°C. Do not freeze. Light-sensitive material. |
For Research Use Only.