missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-17F Monoclonal Antibody (SHLR17), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£166.00 - £335.00
Specifications
| Antigen | IL-17F |
|---|---|
| Clone | SHLR17 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089193
|
Invitrogen™
367-7169-41 |
25 Tests |
£166.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30088548
![]() |
Invitrogen™
367-7169-42 |
100 Tests |
£335.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for Flow CytometrySpecifications
| IL-17F | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| C87042; CANDF6; cytokine ML-1; IL17F; IL-17F; IL-24; ILN; Interleukin; interleukin 17F; Interleukin17F; interleukin-17F; interleukin-24; ML1; ML-1; mutant IL-17F | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| SHLR17 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| Rat | |
| Human | |
| Q96PD4 | |
| 112744 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL17F |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title