Learn More
Invitrogen™ IL-17F (Homodimer) Mouse Uncoated ELISA Kit
Specifically engineered for accurate and precise measurement of mouse IL-17FF homodimer protein levels from samples including serum, and cell culture supernatants.
£174.00 - £392.00
Specifications
Target Species | Mouse |
---|---|
For Use With (Application) | ELISA |
Synonym | Interleukin-17F, IL-17FF |
Alert:
Contains Azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15511157
|
Invitrogen™
88-7472-22 |
2 x 96 Tests |
£174.00
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
15521157
|
Invitrogen™
88-7472-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The Mouse Interleukin 17F (Homodimer) (IL-17F) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-17F. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-17F (Homodimer) Mouse Uncoated ELISA Kit (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)- The assay demonstrates parallelism in measuring recombinant and native mouse IL-17F proteins and a standard curve range of 15 to 2000pg/mL
- No crossreactivity to the IL-17AA homodimer is observed and <2.5% crossreactivity to IL-17AF heterodimer when spiked in excess (100ng/ml)
Specifications
Mouse | |
ELISA | |
Interleukin-17F, IL-17FF |
For Research Use Only