missing translation for 'onlineSavingsMsg'
Learn More

IL-17F Antibody (B-G46) - Azide and BSA Free, Novus Biologicals™

Product Code. 18332566 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332566 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332566 Supplier Novus Biologicals Supplier No. NBP318125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for ELISA (Matched Antibody Pair), ELISPOT
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17F
Applications ELISA
Classification Monoclonal
Clone B-G46
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 1-5ug/ml, ELISPOT 5-10ug/ml
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F
Host Species Mouse
Immunogen Recombinant human IL-17F. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.