missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (B-F60), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14589AF532
This item is not returnable.
View return policy
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for ELISA
Specifications
| IL-17F | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| Recombinant human IL-17F | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA | |
| B-F60 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction